Learn More
Cayman Chemical 3 4MethylendIoxyaPyrrolId 10mg
SDP
Catalog No. 502969899
Encompass_Preferred
An analog of 3,4-MDPV that differs only by the addition of a single carbon to the alkyl chain; intended for forensic and research applications
Catalog No. 50-296-9899 Supplier Cayman Chemical Supplier No. 1636110 MG
An analog of 3,4-MDPV that differs only by the addition of a single carbon to the alkyl chain; intended for forensic and research applications