Learn More
Chemglass Life Sciences 3 and 4mm Bore, 15.2/30 Plug Size, 10mm OD Sidearm
SDP
Catalog No. 501531995
Encompass_Preferred
3 and 4mm Bore, 15.2/30 Plug Size, 10mm OD Sidearm. Glass portion only of CG-424 stopcock. Accepts 1:5 taper PTFE plugs.
Catalog No. 50-153-1995 Supplier Chemglass Life Sciences Supplier No. CGB42605
3 and 4mm Bore, 15.2/30 Plug Size, 10mm OD Sidearm. Glass portion only of CG-424 stopcock. Accepts 1:5 taper PTFE plugs.