Learn More
Wilmad Labglass 3 and 4 mm White Rubber Septa, Precision 100/Pk
Catalog No. NC0987453
Encompass
These Gas-Tight Tube Caps are ideal for experiments that require an air-tight environment under vacuum or low pressure less than 1 bar. Use with our Medium-Walled and Heavy-Walled NMR tubes for better seal and robustness in variable temperature experiments.
Catalog No. NC0987453 Supplier Wilmad Labglass Supplier No. WG3889100
These Gas-Tight Tube Caps are ideal for experiments that require an air-tight environment under vacuum or low pressure less than 1 bar. Use with our Medium-Walled and Heavy-Walled NMR tubes for better seal and robustness in variable temperature experiments.