Learn More
Cayman Chemical 3,4-trimethylene-α-Piperidinovalerophenone (hydrochloride), Purity: ≥98%, 5 mg
SDP
Catalog No. 502916859
Encompass_Preferred
An analytical reference standard categorized as a cathinone; intended for research and forensic applications
Catalog No. 50-291-6859 Supplier Cayman Chemical Supplier No. 377515 MG
An analytical reference standard categorized as a cathinone; intended for research and forensic applications