Learn More
Medchemexpress LLC 3-(5-((((1-(5-(3-chlorophenyl)-3-(trifluoromethyl)pyridin-2-yl)piperidin-4-yl)methyl)amino)methyl)-1-oxoisoindolin-2-yl)piperidine-2,6... | 99.4% | 740.09 | 25 MG
SDP
Catalog No. 5000394298
Encompass_Preferred
MYC degrader 1 TFA (compound A80.2HCl) is an orally available MYC molecular glues degrader with anti-tumor activity. It restores pRB1 protein activity and re-establishes sensitivity of MYC overexpressing cancer cells to CDK4/6 inhibitors.
  • Orally available MYC molecular glues degrader
  • Anti-tumor activity
  • Restores pRB1 protein activity
  • Re-establishes sensitivity of MYC overexpressing cancer cells to CDK4/6 inhibitors
Catalog No. 50-003-94298 Supplier Medchemexpress LLC Supplier No. 25MGHY162318A
MYC degrader 1 TFA (compound A80.2HCl) is an orally available MYC molecular glues degrader with anti-tumor activity. It restores pRB1 protein activity and re-establishes sensitivity of MYC overexpressing cancer cells to CDK4/6 inhibitors.
  • Orally available MYC molecular glues degrader
  • Anti-tumor activity
  • Restores pRB1 protein activity
  • Re-establishes sensitivity of MYC overexpressing cancer cells to CDK4/6 inhibitors