Learn More
Medchemexpress LLC 3-(5-(2-methyl-1H-imidazol-1-yl)-2-(pyrazin-2-ylamino)thiazol-4-yl)benzonitrile | 431040-19-6 | MFCD30747838 | 99.8% | 359.41 | C18H13N7S | 100 MG
SDP
Catalog No. 5000214179
Encompass_Preferred
Adenosine antagonist-1 is a research-grade small-molecule antagonist of the adenosine A3 receptor (AA3R) with reported anti-inflammatory activity. It is supplied as a solid powder and as a 10 mM solution in DMSO for use in biochemical, cellular, and preclinical studies; molecular formula C18H13N7S, molecular weight 359.41, CAS 431040-19-6.
  • Selective adenosine A3 receptor antagonism for receptor biology studies.
  • Reported anti-inflammatory activity in preclinical models.
  • Available as solid powder or 10 mM solution in DMSO for flexible dosing.
  • High purity suitable for biochemical and cellular assays.
  • Molecular formula and weight provided for formulation and dosing calculations.
Catalog No. 50-002-14179 Supplier Medchemexpress LLC Supplier No. HY100274100MG
Adenosine antagonist-1 is a research-grade small-molecule antagonist of the adenosine A3 receptor (AA3R) with reported anti-inflammatory activity. It is supplied as a solid powder and as a 10 mM solution in DMSO for use in biochemical, cellular, and preclinical studies; molecular formula C18H13N7S, molecular weight 359.41, CAS 431040-19-6.
  • Selective adenosine A3 receptor antagonism for receptor biology studies.
  • Reported anti-inflammatory activity in preclinical models.
  • Available as solid powder or 10 mM solution in DMSO for flexible dosing.
  • High purity suitable for biochemical and cellular assays.
  • Molecular formula and weight provided for formulation and dosing calculations.