Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3 5 3 -Triiodo-D-thy 1mg | 1393257 | 650.97 g/mol | C15H12I3NO4 | 1 MG
SDP

Catalog No. 5000226561
Encompass_Preferred

3,5,3'-Triiodo-D-thyronine (D-T3) is the D-isomer of the thyroid hormone triiodothyronine supplied as a research reagent. It is reported to be less effective than the L-isomer (T3) in normalizing increased plasma creatine phosphokinase (CPK) activity. Product is offered in small research pack sizes for laboratory use.

  • Research reagent for thyroid hormone and metabolic studies.
  • D-isomer of triiodothyronine with lower biological potency than the L-isomer.
  • CAS number 5714-08-9; molecular formula C15H12I3NO4.
  • Molar mass approximately 650.97 g/mol.
  • Typical small pack size: 1 mg for laboratory use.

Catalog No. 50-002-26561 Supplier Medchemexpress LLC Supplier No. HY1163251MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3,5,3'-Triiodo-D-thyronine (D-T3) is the D-isomer of the thyroid hormone triiodothyronine supplied as a research reagent. It is reported to be less effective than the L-isomer (T3) in normalizing increased plasma creatine phosphokinase (CPK) activity. Product is offered in small research pack sizes for laboratory use.

  • Research reagent for thyroid hormone and metabolic studies.
  • D-isomer of triiodothyronine with lower biological potency than the L-isomer.
  • CAS number 5714-08-9; molecular formula C15H12I3NO4.
  • Molar mass approximately 650.97 g/mol.
  • Typical small pack size: 1 mg for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.