Learn More
Medchemexpress LLC 3-[5-[4-(2-hydroxy-2-methylpropanoyl)piperazin-1-yl]-2-(trifluoromethyl)phenyl]-4-(1H-indol-3-yl)pyr | 1260181-14-3 | MFCD19380092 | 99.5% | 526.51 | C27H25F3N4O4 | 10 MG
SDP
Catalog No. 5000242671
Encompass_Preferred
TCS 21311 is a small-molecule research compound that potently and selectively inhibits JAK3 and exhibits additional activity against GSK-3 and PKC isoforms, intended for in vitro and preclinical research use.
  • Potent JAK3 inhibition (IC50 8 nM).
  • Secondary activity against GSK-3β and PKC isoforms (IC50s 3-68 nM).
  • High chemical purity (~99.5%).
  • Molecular weight 526.51 g/mol; formula C27H25F3N4O4.
  • Available in small-mass formats and DMSO solution for ready reconstitution.
Catalog No. 50-002-42671 Supplier Medchemexpress LLC Supplier No. HY10826410MG
TCS 21311 is a small-molecule research compound that potently and selectively inhibits JAK3 and exhibits additional activity against GSK-3 and PKC isoforms, intended for in vitro and preclinical research use.
  • Potent JAK3 inhibition (IC50 8 nM).
  • Secondary activity against GSK-3β and PKC isoforms (IC50s 3-68 nM).
  • High chemical purity (~99.5%).
  • Molecular weight 526.51 g/mol; formula C27H25F3N4O4.
  • Available in small-mass formats and DMSO solution for ready reconstitution.