Learn More
Medchemexpress LLC 3-[5-[4-(2-hydroxy-2-methylpropanoyl)piperazin-1-yl]-2-(trifluoromethyl)phenyl]-4-(1H-indol-3-yl)pyrrole-2,5-dione | 1260181-14-3 | MFCD19380092 | 99.5% | 526.51 g/mol | C27H25F3N4O4 | 5 MG
SDP
Catalog No. 5000243006
Encompass_Preferred
TCS 21311 (NIBR3049) is a research-stage small molecule that potently and selectively inhibits Janus kinase 3 (JAK3). It is used in biochemical and cellular studies to probe JAK3-dependent signaling pathways and has reported activity against PKC and GSK-3β.
  • Selective JAK3 inhibitor with IC50 approximately 8 nM.
  • Also inhibits PKC and GSK-3β, useful for kinase profiling.
  • High purity (99.5%) suitable for biochemical assays.
  • Solid form, white to yellow, with defined storage stability.
  • Supplied in small milligram quantities for research applications.
Catalog No. 50-002-43006 Supplier Medchemexpress LLC Supplier No. HY1082645MG
TCS 21311 (NIBR3049) is a research-stage small molecule that potently and selectively inhibits Janus kinase 3 (JAK3). It is used in biochemical and cellular studies to probe JAK3-dependent signaling pathways and has reported activity against PKC and GSK-3β.
  • Selective JAK3 inhibitor with IC50 approximately 8 nM.
  • Also inhibits PKC and GSK-3β, useful for kinase profiling.
  • High purity (99.5%) suitable for biochemical assays.
  • Solid form, white to yellow, with defined storage stability.
  • Supplied in small milligram quantities for research applications.