Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5,5-Trimethylhexyl Acetate 93.0+%, TCI America™
SDP

Catalog No. A098225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A098225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A098225ML Supplier TCI America Supplier No. A098225ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 58430-94-7
Color Colorless
Boiling Point 209°C
Molecular Formula C11H22O2
Quantity 25 mL
MDL Number MFCD00026336
Synonym Acetic Acid 3,5,5-Trimethylhexyl Ester
InChI Key DGKXDLCVQSQVBC-UHFFFAOYSA-N
SMILES CC(CCOC(=O)C)CC(C)(C)C
IUPAC Name 3,5,5-trimethylhexyl acetate
Molecular Weight (g/mol) 186.295
PubChem CID 42745
Formula Weight 186.30
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,5,5-Trimethylhexyl Acetate
Show More Show Less
EINECSNumber (2)-0735
RTECSNumber AK0840000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.