Learn More
Cayman Chemical 3,5,6,7,8,3',4'-Heptamethoxyflavone, Purity: ≥98%, CAS: 1178-24-1, 5 mg
SDP
Catalog No. 502911862
Encompass_Preferred
A polymethoxyflavone; inhibits the efflux of Hoechst 33342 by BCRP and rhodamine 123 by P-gp in MDCK cells expressing the human transporters (IC50s = 1.4 and 31 µM, respectively); prevents UVB-induced decreases in type I procollagen in HDFn cells at 50, 100, and 200 µg/ml; inhibits the proliferation of B16 melanoma 4A5, CCRF-HSB-2 leukemia, and TGBC11TKB gastric cancer cells (IC50s = 38, 29, and 9 µM, respectively); prevents corticosterone-induced increases in immobility time in the forced swim test in mice at 50 mg/kg
Catalog No. 50-291-1862 Supplier Cayman Chemical Supplier No. 364755 MG
A polymethoxyflavone; inhibits the efflux of Hoechst 33342 by BCRP and rhodamine 123 by P-gp in MDCK cells expressing the human transporters (IC50s = 1.4 and 31 µM, respectively); prevents UVB-induced decreases in type I procollagen in HDFn cells at 50, 100, and 200 µg/ml; inhibits the proliferation of B16 melanoma 4A5, CCRF-HSB-2 leukemia, and TGBC11TKB gastric cancer cells (IC50s = 38, 29, and 9 µM, respectively); prevents corticosterone-induced increases in immobility time in the forced swim test in mice at 50 mg/kg