Learn More
Medchemexpress LLC (3α,5β,6β,7α)-BAR501 | 1632118-69-4 | 99.9% | 406.64 g·mol⁻¹ | C26H46O3 | 1 ML
SDP
Catalog No. 5000211694
Encompass_Preferred
A cholane-scaffold small molecule that functions as a selective agonist of GPBAR1 (TGR5) and is used in research on FXR and bile-acid receptor-mediated signaling. Supplied in solution and solid formats for flexibility in assay setup, it is intended for in vitro pharmacology and signaling studies.
  • High purity suitable for biochemical and cell-based assays.
  • Available as a 10 mM solution in DMSO for immediate use in assays.
  • Also offered as solid material in multiple sizes for dose optimization.
  • Molecular formula and weight provided to support analytical planning.
  • Commonly used in studies of bile acid receptor signaling and metabolic disease models.
Catalog No. 50-002-11694 Supplier Medchemexpress LLC Supplier No. HY101274A10MM 1ML
A cholane-scaffold small molecule that functions as a selective agonist of GPBAR1 (TGR5) and is used in research on FXR and bile-acid receptor-mediated signaling. Supplied in solution and solid formats for flexibility in assay setup, it is intended for in vitro pharmacology and signaling studies.
  • High purity suitable for biochemical and cell-based assays.
  • Available as a 10 mM solution in DMSO for immediate use in assays.
  • Also offered as solid material in multiple sizes for dose optimization.
  • Molecular formula and weight provided to support analytical planning.
  • Commonly used in studies of bile acid receptor signaling and metabolic disease models.