Learn More
Medchemexpress LLC 3-[[[5-(6-Methyl-2-pyridinyl)-4-triazolo[1,5-a]pyridin-6-yl-1H-imidazol-2-yl]methyl]amino]benzonitrile | 1352609-28-9 | 98.8% | 406.44 | C23H18N8 | 10 MG
SDP
Catalog No. 5000299221
Encompass_Preferred
EW-7195 is a potent, selective ALK5 (TGFβR1) inhibitor for research use that blocks TGF-β1-induced Smad signaling, epithelial-to-mesenchymal transition, and metastatic progression in preclinical studies. It provides low-nanomolar potency and is supplied with high purity for laboratory assays.
  • Potent ALK5 inhibition with low-nanomolar IC50.
  • High selectivity over related kinases, >300-fold versus p38α.
  • Blocks TGF-β1-induced Smad signaling and epithelial-to-mesenchymal transition.
  • Demonstrated anti-metastasis activity in preclinical models.
  • High purity suitable for research applications.
  • Soluble in DMSO for assay preparation.
Catalog No. 50-002-99221 Supplier Medchemexpress LLC Supplier No. HY1876610MG
EW-7195 is a potent, selective ALK5 (TGFβR1) inhibitor for research use that blocks TGF-β1-induced Smad signaling, epithelial-to-mesenchymal transition, and metastatic progression in preclinical studies. It provides low-nanomolar potency and is supplied with high purity for laboratory assays.
  • Potent ALK5 inhibition with low-nanomolar IC50.
  • High selectivity over related kinases, >300-fold versus p38α.
  • Blocks TGF-β1-induced Smad signaling and epithelial-to-mesenchymal transition.
  • Demonstrated anti-metastasis activity in preclinical models.
  • High purity suitable for research applications.
  • Soluble in DMSO for assay preparation.