Learn More
Medchemexpress LLC 3,5,6-trichloro-2-pyridinol | 6515-38-4 | MFCD00130269 | 99.9% | 198.43 g·mol⁻¹ | C5H2Cl3NO | 1 ML
SDP
Catalog No. 5000301628
Encompass_Preferred
3,5,6-Trichloro-2-pyridinol is supplied as a 10 mM solution in DMSO (1 mL) for research and analytical use. The compound has CAS 6515-38-4 and a molecular weight of 198.43 g·mol⁻¹. Follow recommended storage guidelines for solution and powder to maintain stability.
  • High purity (99.85%).
  • Ready-to-use 10 mM solution in DMSO.
  • Compact 1 mL vial for small-scale assays.
  • Suitable for analytical and research applications.
  • Recommended storage: in solvent -80°C (6 months) or -20°C (1 month).
Catalog No. 50-003-01628 Supplier Medchemexpress LLC Supplier No. HYW01817110MM/1ML
3,5,6-Trichloro-2-pyridinol is supplied as a 10 mM solution in DMSO (1 mL) for research and analytical use. The compound has CAS 6515-38-4 and a molecular weight of 198.43 g·mol⁻¹. Follow recommended storage guidelines for solution and powder to maintain stability.
  • High purity (99.85%).
  • Ready-to-use 10 mM solution in DMSO.
  • Compact 1 mL vial for small-scale assays.
  • Suitable for analytical and research applications.
  • Recommended storage: in solvent -80°C (6 months) or -20°C (1 month).