Learn More
Medchemexpress LLC 3,5,6-trichloro-2-pyridinol | 6515-38-4 | MFCD00130269 | 99.9% | 198.43 g·mol⁻¹ | C5H2Cl3NO | 5 G
SDP
Catalog No. 5000299685
Encompass_Preferred
3,5,6-Trichloro-2-pyridinol is a chlorinated pyridinol compound and a primary degradation metabolite of certain organophosphate insecticides and herbicides. It is provided for laboratory use as an analytical standard and for toxicology, environmental monitoring, and research applications.
  • High purity suitable for analytical and toxicology studies.
  • Available as solid in multiple gram sizes and as solution formats.
  • Molecular formula C5H2Cl3NO; molecular weight 198.43 g·mol⁻¹.
  • Characterized by LC-MS with a certificate of analysis for each batch.
  • Suitable as a metabolite reference and environmental marker.
Catalog No. 50-002-99685 Supplier Medchemexpress LLC Supplier No. HYW0181715G
3,5,6-Trichloro-2-pyridinol is a chlorinated pyridinol compound and a primary degradation metabolite of certain organophosphate insecticides and herbicides. It is provided for laboratory use as an analytical standard and for toxicology, environmental monitoring, and research applications.
  • High purity suitable for analytical and toxicology studies.
  • Available as solid in multiple gram sizes and as solution formats.
  • Molecular formula C5H2Cl3NO; molecular weight 198.43 g·mol⁻¹.
  • Characterized by LC-MS with a certificate of analysis for each batch.
  • Suitable as a metabolite reference and environmental marker.