Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,5,9-trioxa-4-phosphaheptacosan-1-aminium, 4,7-dihydroxy-N,N,N-trimethyl-, inner salt, 4-oxide | 74430-89-0 | MFCD00065889 | 99.7% | C26H56NO6P | 10MG
SDP

Catalog No. 5000258920
Encompass_Preferred

Lyso-PAF C-18 is a research-grade chemical intermediate used in the synthesis of C18-PAF, a ligand for the platelet-activating factor receptor. It is supplied as a white to off-white solid with high reported purity and is intended for laboratory research use only.

  • High purity (99.7%).
  • White to off-white solid form.
  • Molecular formula C26H56NO6P.
  • Molecular weight 509.70 g/mol.
  • Powder stable at -20°C for up to 3 years.
  • In-solution storage: -80°C for 6 months, -20°C for 1 month.
  • Supplied in small research sizes suitable for synthesis and analytical work.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-58920 Supplier Medchemexpress LLC Supplier No. HY14158110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lyso-PAF C-18 is a research-grade chemical intermediate used in the synthesis of C18-PAF, a ligand for the platelet-activating factor receptor. It is supplied as a white to off-white solid with high reported purity and is intended for laboratory research use only.

  • High purity (99.7%).
  • White to off-white solid form.
  • Molecular formula C26H56NO6P.
  • Molecular weight 509.70 g/mol.
  • Powder stable at -20°C for up to 3 years.
  • In-solution storage: -80°C for 6 months, -20°C for 1 month.
  • Supplied in small research sizes suitable for synthesis and analytical work.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.