Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Bis(3,5-dimethoxybenzyloxy)benzyl Bromide 97.0+%, TCI America™
SDP

Catalog No. B22471G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22471G 1 g
B22475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B22471G Supplier TCI America Supplier No. B22471G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 176650-93-4
Molecular Formula C25H27BrO6
Molecular Weight (g/mol) 503.39
MDL Number MFCD04038410
InChI Key RLACLNXCSSBIKS-UHFFFAOYSA-N
PubChem CID 15778113
IUPAC Name 1-{[3-(bromomethyl)-5-[(3,5-dimethoxyphenyl)methoxy]phenoxy]methyl}-3,5-dimethoxybenzene
SMILES COC1=CC(OC)=CC(COC2=CC(OCC3=CC(OC)=CC(OC)=C3)=CC(CBr)=C2)=C1

Specifications

Quantity 1 g
Formula Weight 503.39
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Bis(3,5-dimethoxybenzyloxy)benzyl Bromide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.