Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Bis(methoxymethyloxy)benzyl Alcohol 98.0+%, TCI America™
SDP

Catalog No. B55831G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B55831G 1 g
B55835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B55831G Supplier TCI America Supplier No. B55831G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 76280-60-9
Molecular Formula C11H16O5
Molecular Weight (g/mol) 228.24
MDL Number MFCD27928465
InChI Key QZORVGYVDDOQNT-UHFFFAOYSA-N
Synonym [3,5-Bis(methoxymethoxy)phenyl]methanol
PubChem CID 10443751
IUPAC Name [3,5-bis(methoxymethoxy)phenyl]methanol
SMILES COCOC1=CC(OCOC)=CC(CO)=C1

Specifications

Color Yellow
Boiling Point 172°C
Quantity 1 g
Formula Weight 228.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,5-Bis(methoxymethyloxy)benzyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.