Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Bis(tert-butyldiphenylsilyloxy)benzyl Alcohol 95.0+%, TCI America™
SDP

Catalog No. B20521G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B20521G Supplier TCI America Supplier No. B20521G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 182250-70-0
Molecular Formula C39H44O3Si2
Molecular Weight (g/mol) 616.948
MDL Number MFCD02093440
InChI Key SDDISUZNVYXXHS-UHFFFAOYSA-N
PubChem CID 11114774
IUPAC Name [3,5-bis[[tert-butyl(diphenyl)silyl]oxy]phenyl]methanol
SMILES CC(C)(C)[Si](C1=CC=CC=C1)(C2=CC=CC=C2)OC3=CC(=CC(=C3)CO)O[Si](C4=CC=CC=C4)(C5=CC=CC=C5)C(C)(C)C

Specifications

Melting Point 108°C
Color White
Quantity 1 g
Formula Weight 616.95
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Bis(tert-butyldiphenylsilyloxy)benzyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.