Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Bis(trifluoromethyl)-2-chloroaniline 98.0+%, TCI America™
SDP

Catalog No. B277725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B277725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B277725G Supplier TCI America Supplier No. B277725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 201593-90-0
Molecular Formula C8H4ClF6N
Molecular Weight (g/mol) 263.567
InChI Key JKUFETFGEJPHEM-UHFFFAOYSA-N
Synonym 2-chloro-3,5-bis trifluoromethyl aniline, 3,5-bis trifluoromethyl-2-chloroaniline, 2-chloro-3,5-bis-trifluoromethyl aniline, benzenamine, 2-chloro-3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl-2-chlorophenylamine, pubchem2938, 2-chloro-3,5-bis-trifluoromethyl-aniline, 3,5-bis trifluoromethyl-2-chloro aniline, 2-chloranyl-3,5-bis trifluoromethyl aniline, 2-chloro-3,5-bis-trifluoromethyl-phenylamine
PubChem CID 2757429
IUPAC Name 2-chloro-3,5-bis(trifluoromethyl)aniline
SMILES C1=C(C=C(C(=C1C(F)(F)F)Cl)N)C(F)(F)F

Specifications

Color Yellow
Quantity 25 g
Formula Weight 263.57
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,5-Bis(trifluoromethyl)-2-chloroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.