Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Bis(trifluoromethyl)benzamide 98.0+%, TCI America™
SDP

Catalog No. B18825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B18825G Supplier TCI America Supplier No. B18825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 22227-26-5
Molecular Formula C9H5F6NO
Molecular Weight (g/mol) 257.135
MDL Number MFCD00015511
InChI Key YNOPIKHMZIOWHS-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzamide, 3,5-di trifluoromethyl benzamide, 3,5-bis trifluoromethyl-benzamide, benzamide, 3,5-bis trifluoromethyl, acmc-1cglb, ynopikhmziowhs-uhfffaoysa, 3,5-bis-trifluoromethyl-benzamide, 3,5-bis-trifluoromethyl benzamide, benzamide,3,5-bis trifluoromethyl
PubChem CID 519927
IUPAC Name 3,5-bis(trifluoromethyl)benzamide
SMILES C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C(=O)N

Specifications

Melting Point 164°C
Color White
Quantity 5 g
Formula Weight 257.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Bis(trifluoromethyl)benzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.