Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Bis(trifluoromethyl)phenol, 97%

Catalog No. AAA1018306
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1018306 5 g
AAA1018303 1 g
AAA1018314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1018306 Supplier Thermo Scientific Chemicals Supplier No. A1018306
Only null left
Add to Cart

CAS: 349-58-6 | C8H4F6O | 230.109 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 349-58-6
Molecular Formula C8H4F6O
Molecular Weight (g/mol) 230.109
MDL Number MFCD00000386
InChI Key ODSXJQYJADZFJX-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl phenol, phenol, 3,5-bis trifluoromethyl, 3,5-di trifluoromethyl phenol, 3,5-bis-trifluoromethylphenol, 3,5-ditrifluoromethylphenol, 3,5-bis-trifluoromethyl phenol, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-3,5-xylenol, pubchem12483, mbt-oh, acmc-1clsr
PubChem CID 67680
IUPAC Name 3,5-bis(trifluoromethyl)phenol
SMILES C1=C(C=C(C=C1C(F)(F)F)O)C(F)(F)F

Specifications

Melting Point 20°C to 21°C
Density 1.511
Boiling Point 97°C to 98°C (50 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.415
Quantity 5 g
Beilstein 2123670
Formula Weight 230.11
Percent Purity 97%
Chemical Name or Material 3,5-Bis (trifluoromethyl)phenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-488-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.