Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,5-Bis(trifluoromethyl)phenyl isothiocyanate

Catalog No. 4685171G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-851-71G 1 g
46-851-75G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 46-851-71G Supplier Sigma Aldrich Supplier No. 4685171G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3,5-bis(trifluoromethyl)phenyl isothiocyanate has been employed for the following studies:
  • Chemical derivatization of amino-functionalized model surfaces, amino thiolate on au, amino siloxane on si, and polyethylene (pe) foils and films
  • Chemical derivatization of self assembled monolayers of -d-amino-4,4α-terphenyl substituted alkanethiols
  • Synthesis of ethylene-linked sulfonimidoyl-containing thioureas
  • Preparation of pyrrolidine-2-carboxylic acid-{2-[3-(3,5-bistrifluoromethylphenyl)thioureido]phenyl}-amide

Specifications

CAS 23165-29-9
Density 1.485 g/mL (at 25°C (literature))
Boiling Point 63°C (1.5 mmHg)
Molecular Formula C9H3F6NS
Refractive Index n20/D 1.5 (literature)
Linear Formula (CF3)2C6H3NCS
MDL Number MFCD00040838
Quantity 1 g
Molecular Weight (g/mol) 271.18
Percent Purity 98%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.