Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC [(3,5-bis(trifluoromethyl)phenyl)methyl] azabicyclohexane carboxylate | 2162961-71-7 | >98.0% | 527.4 g/mol | C23H19F6N5O3 | 10MM 1ML
SDP

Catalog No. 5000236136
Encompass_Preferred

BI-2545 is a potent small-molecule autotaxin (ATX) inhibitor used as a preclinical chemical probe to reduce lysophosphatidic acid (LPA) levels in vitro and in vivo. It exhibits single-digit nanomolar enzyme potency and measurable whole-blood activity; supplied for research use as a solid or concentrated solution. CAS: 2162961-71-7. Chemical formula: C23H19F6N5O3.

  • Potent ATX inhibition in enzyme assays (human ATX IC50 2.2 nM; rat ATX IC50 3.4 nM).
  • Demonstrated whole-blood activity (human IC50 29 nM; rat IC50 96 nM).
  • Suitable for in vitro and in vivo preclinical studies.
  • High purity suitable for research (>98.0%).
  • Molecular weight 527.4 g/mol; chemical formula C23H19F6N5O3.
  • Available as solid or concentrated solution for dosing flexibility.

Catalog No. 50-002-36136 Supplier Medchemexpress LLC Supplier No. HY12477210MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BI-2545 is a potent small-molecule autotaxin (ATX) inhibitor used as a preclinical chemical probe to reduce lysophosphatidic acid (LPA) levels in vitro and in vivo. It exhibits single-digit nanomolar enzyme potency and measurable whole-blood activity; supplied for research use as a solid or concentrated solution. CAS: 2162961-71-7. Chemical formula: C23H19F6N5O3.

  • Potent ATX inhibition in enzyme assays (human ATX IC50 2.2 nM; rat ATX IC50 3.4 nM).
  • Demonstrated whole-blood activity (human IC50 29 nM; rat IC50 96 nM).
  • Suitable for in vitro and in vivo preclinical studies.
  • High purity suitable for research (>98.0%).
  • Molecular weight 527.4 g/mol; chemical formula C23H19F6N5O3.
  • Available as solid or concentrated solution for dosing flexibility.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.