Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Bis(trifluoromethyl)phenylacetic acid, 97%

Catalog No. AAA1892206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1892206 5 g
AAA1892203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1892206 Supplier Thermo Scientific Chemicals Supplier No. A1892206
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 85068-33-3 | C10H6F6O2 | 272.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85068-33-3
Molecular Formula C10H6F6O2
Molecular Weight (g/mol) 272.15
MDL Number MFCD00009908
InChI Key PAWSKKHEEYTXSA-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl phenylacetic acid, 2-3,5-bis trifluoromethyl phenyl acetic acid, 3,5-bis-trifluoromethylphenylacetic acid, mbt-paa, 2-3,5-di trifluoromethyl phenyl acetic acid, benzeneacetic acid, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl phenyl acetic acid, 3,5-di trifluoromethyl phenylacetic acid, 3,5-bis trifluoromethyl phenylaceticacid
PubChem CID 123587
IUPAC Name 2-[3,5-bis(trifluoromethyl)phenyl]acetic acid
SMILES OC(=O)CC1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Specifications

Melting Point 121°C to 123°C
Quantity 5 g
Beilstein 6813447
Formula Weight 272.15
Percent Purity 97%
Chemical Name or Material 3,5-Bis(trifluoromethyl)phenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 285-295-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.