Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Bis(trifluoromethyl)phenylacetonitrile 98.0+%, TCI America™
SDP

Catalog No. B29115G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B29115G Supplier TCI America Supplier No. B29115G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 85068-32-2
Molecular Formula C10H5F6N
Molecular Weight (g/mol) 253.15
MDL Number MFCD00009904
InChI Key YXGWYBUKRTYHJM-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl phenylacetonitrile, 2-3,5-bis trifluoromethyl phenyl acetonitrile, 3,5-bis trifluoromethyl benzyl cyanide, 3,5-di trifluoromethyl benzyl cyanide, 3,5-bis trifluoromethyl-phenylacetonitrile, 3,5-bis trifluoromethyl phenyl acetonitrile, benzeneacetonitrile, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl benzylcyanide
PubChem CID 603657
IUPAC Name 2-[3,5-bis(trifluoromethyl)phenyl]acetonitrile
SMILES FC(F)(F)C1=CC(=CC(CC#N)=C1)C(F)(F)F

Specifications

Color Yellow
Quantity 5 g
UN Number 3276
Formula Weight 253.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,5-Bis(trifluoromethyl)phenylacetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.