Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Bis(trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. B18865G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1886-5G 5 g
B1886-25G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1886-5G Supplier TCI America Supplier No. B18865G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 73852-19-4
Melting Point 220°C
Color White-Yellow
Molecular Formula C8H5BF6O2
Quantity 5 g
MDL Number MFCD00051850
Synonym 3,5-bis trifluoromethyl phenylboronic acid, 3,5-bis trifluoromethyl benzeneboronic acid, 3,5-bis trifluoromethyl phenyl boronic acid, 3,5-bis-trifluoromethylphenylboronic acid, 3,5-bis trifluoromethylphenyl boronic acid
InChI Key BPTABBGLHGBJQR-UHFFFAOYSA-N
SMILES B(C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)(O)O
IUPAC Name [3,5-bis(trifluoromethyl)phenyl]boronic acid
Molecular Weight (g/mol) 257.926
PubChem CID 156265
Formula Weight 257.93
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.