Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Bis(trifluoromethyl)pyrazole, 97%, Thermo Scientific™

Catalog No. AC311810010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC311810010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC311810010 Supplier Thermo Scientific Chemicals Supplier No. 311810010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14704-41-7
Molecular Formula C5H2F6N2
Molecular Weight (g/mol) 204.07
InChI Key NGDDUAYSWPUSLX-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl-1h-pyrazole, 3,5-bis trifluoromethyl pyrazole, 1h-pyrazole,3,5-bis trifluoromethyl, 1h-pyrazole, 3,5-bis trifluoromethyl, 3,3-bis trifluoromethyl pyrazol, 3,5-bis trifluoromethyl pyrazol, 3,5-bis-trifluoromethyl-1h-pyrazole, bis trifluoromethyl pyrazole, 3,5-di trifluoromethyl-1h-pyrazole
PubChem CID 518991
IUPAC Name 3,5-bis(trifluoromethyl)-1H-pyrazole
SMILES C1=C(NN=C1C(F)(F)F)C(F)(F)F

Specifications

Melting Point 83.0°C to 85.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 204.07
Percent Purity 97%
Chemical Name or Material 3,5-Bis(trifluoromethyl)pyrazole
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.