Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Bis(trifluoromethyl)pyrazole 98.0+%, TCI America™
SDP

Catalog No. B47625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47625G 5 g
B47621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47625G Supplier TCI America Supplier No. B47625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14704-41-7
Molecular Formula C5H2F6N2
Molecular Weight (g/mol) 204.075
MDL Number MFCD00153672
InChI Key NGDDUAYSWPUSLX-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl-1h-pyrazole, 3,5-bis trifluoromethyl pyrazole, 1h-pyrazole,3,5-bis trifluoromethyl, 1h-pyrazole, 3,5-bis trifluoromethyl, 3,3-bis trifluoromethyl pyrazol, 3,5-bis trifluoromethyl pyrazol, 3,5-bis-trifluoromethyl-1h-pyrazole, bis trifluoromethyl pyrazole, 3,5-di trifluoromethyl-1h-pyrazole
PubChem CID 518991
IUPAC Name 3,5-bis(trifluoromethyl)-1H-pyrazole
SMILES C1=C(NN=C1C(F)(F)F)C(F)(F)F

Specifications

Melting Point 85°C
Color White-Yellow
Boiling Point 147°C
Quantity 5 g
Formula Weight 204.08
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Bis(trifluoromethyl)pyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.