Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Bis(trifluoromethyl)styrene (stabilized with TBC) 98.0+%, TCI America™
SDP

Catalog No. B37425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B37425G Supplier TCI America Supplier No. B37425G

Specifications

CAS 349-59-7
Melting Point 8°C
Color Yellow
Molecular Formula C10H6F6
MDL Number MFCD00075538
UN Number 1993
Quantity 5 g
Synonym 3,5-bis trifluoromethyl styrene, 1,3-bis trifluoromethyl-5-vinylbenzene, 1-ethenyl-3,5-bis trifluoromethyl benzene, 3,5-bis trifluoromethyl styrene, stab. with 4-methoxyphenol, pubchem2736, acmc-1cmtq, 3,5-bis trifluromethyl styrene, 1,3-bis-trifluoromethyl-5-vinyl-benzene, 3,5-bis trifluoromethyl styrene, stab.
InChI Key LFICVUCVPKKPFF-UHFFFAOYSA-N
SMILES FC(F)(F)C1=CC(=CC(C=C)=C1)C(F)(F)F
IUPAC Name 1-ethenyl-3,5-bis(trifluoromethyl)benzene
Molecular Weight (g/mol) 240.15
PubChem CID 2773242
Formula Weight 240.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,5-Bis(trifluoromethyl)styrene (stabilized with TBC)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.