Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,5-Bis(trifluoromethyl)toluene

Catalog No. MNO0001481G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MNO0001481G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MNO0001481G Supplier Sigma Aldrich Supplier No. MNO0001481G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • N-boc-pyrrolidine is an n-substituted pyrrolidine. it is reported that the reactivity of n-boc-pyrrolidine towards c-h insertion reaction is 2000 times more than cyclohexane. it undergoes α-arylation in the presence of a palladium catalyst with high enantioselectivity.n-boc-pyrrolidine may be used in the synthesis of the following:
  • 2-aryl-n-boc-pyrrolidines
  • Scalemic 2-pyrrolidinylcuprates
  • 2-alkenyl-n-boc-pyrrolidines
  • 1-deoxycastanospermine
  • Methylphenidate analogues
  • (+)-elaeokanine a

Specifications

CAS 75462-61-2
Molecular Formula C9H6F6
Linear Formula C9H6F6
Quantity 10 g
MDL Number MFCD04972861
Molecular Weight (g/mol) 228.13
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.