Learn More
Medchemexpress LLC 3',5'-cyclic guanosine monophosphate | 7665-99-8 | MFCD00070129 | 99.8% | 345.21 | C10H12N5O7P | 5 MG
SDP
Catalog No. 5000263771
Encompass_Preferred
Cyclic GMP (cGMP) is a cyclic nucleotide that serves as an intracellular second messenger mediating responses to nitric oxide and natriuretic peptides. This research-grade reagent is provided as a high-purity analytical standard for biochemical assays, signaling pathway studies, and reference applications.
  • High purity suitable for analytical and biochemical work.
  • Functions as an intracellular second messenger in signaling pathways.
  • Compatible with assay standards and reference materials.
  • Molecular weight 345.21 and formula C10H12N5O7P.
  • Available in milligram-scale quantities for small-scale experiments.
Catalog No. 50-002-63771 Supplier Medchemexpress LLC Supplier No. HY1134695MG
Cyclic GMP (cGMP) is a cyclic nucleotide that serves as an intracellular second messenger mediating responses to nitric oxide and natriuretic peptides. This research-grade reagent is provided as a high-purity analytical standard for biochemical assays, signaling pathway studies, and reference applications.
  • High purity suitable for analytical and biochemical work.
  • Functions as an intracellular second messenger in signaling pathways.
  • Compatible with assay standards and reference materials.
  • Molecular weight 345.21 and formula C10H12N5O7P.
  • Available in milligram-scale quantities for small-scale experiments.