Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Di(2-pyridyl)pyrazole 98.0+%, TCI America™
SDP

Catalog No. D20375G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D20375G 5 g
D20371G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D20375G Supplier TCI America Supplier No. D20375G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 129485-83-2
Molecular Formula C13H10N4
Molecular Weight (g/mol) 222.25
MDL Number MFCD00130125
InChI Key IMDRKCUYKQQEAC-UHFFFAOYSA-N
PubChem CID 613762
IUPAC Name 2-[3-(pyridin-2-yl)-1H-pyrazol-5-yl]pyridine
SMILES N1N=C(C=C1C1=CC=CC=N1)C1=CC=CC=N1

Specifications

Melting Point 188°C
Color White-Yellow
Quantity 5 g
Formula Weight 222.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Di(2-pyridyl)pyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.