Learn More
Enzo Life Sciences 3,5-Di-O-caffeoylquinic acid (1mg). CAS: 2450-53-5
SDP
Supplier:  Enzo Life Sciences ALX350320M001
Encompass_Preferred
Antioxidant. Shows antiproliferative activity. Alternative name: 3,5-CQA, Isochlorogenic acid. Purity: ≥97% (HPLC). Appearance: Off-white to yellow/brown crystalline solid. Solubility: Soluble in DMSO or 100% ethanol. Long Term Storage: -20°C.
Catalog No. 50-200-8842