Learn More
Medchemexpress LLC 3,5-Di-tert-butyl-4-hydroxyphenylpropionic acid | 20170-32-5 | 99.95% | 278.39 | 10 G

Supplier: Medchemexpress LLC HYW00934710G
COX-1-IN-3 is identified as a COX-1 inhibitor with non-steroidal anti-inflammatory activity. It is intended for research use only.
- Inhibits COX-1.
- Exhibits non-steroidal anti-inflammatory activity.
- Purity of 99.95%.
- Available in solid and solution forms.
- Can be stored at room temperature for 3 years (solid form).
- In solvent, can be stored at -80°C for 2 years or -20°C for 1 year.
- Soluble in DMSO (100 mg/mL).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.