Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Di-tert-butylbenzaldehyde 97.0+%, TCI America™
SDP

Catalog No. D41301G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41301G 1 g
D41305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41301G Supplier TCI America Supplier No. D41301G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17610-00-3
Molecular Formula C15H22O
Molecular Weight (g/mol) 218.34
MDL Number MFCD00026298
InChI Key BRUITYMDHWNCIG-UHFFFAOYSA-N
Synonym 3,5-di-tert-butylbenzaldehyde, 3,5-bis tert-butyl benzaldehyde, 3,5-di-t-butylbenzaldehyde, 3,5-bis tert-butyl-1-formylbenzene, benzaldehyde, 3,5-bis 1,1-dimethylethyl, 3,5-di-tert-butyl-benzaldehyde, acmc-20ai73, 3,5-di-tertbutylbenzaldehyde, ksc181g1d, 3,5-di-tert butylbenzaldehyde
PubChem CID 1268253
IUPAC Name 3,5-di-tert-butylbenzaldehyde
SMILES CC(C)(C)C1=CC(=CC(C=O)=C1)C(C)(C)C

Specifications

Melting Point 86°C
Color White
Boiling Point 132°C
Quantity 1 g
Formula Weight 218.34
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Di-tert-butylbenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.