Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Di-tert-butylcatechol, 99%

Catalog No. AAH5520714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5520714 25 g
AAH5520706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5520714 Supplier Thermo Scientific Chemicals Supplier No. H5520714
Only null left
Add to Cart

CAS: 1020-31-1 | C14H22O2 | 222.328 g/mol

It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Insoluble in water.

Notes
Store at room temperature with proper ventilation. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1020-31-1
Molecular Formula C14H22O2
Molecular Weight (g/mol) 222.328
MDL Number MFCD00008819
InChI Key PJZLSMMERMMQBJ-UHFFFAOYSA-N
Synonym 3,5-di-tert-butylcatechol, 3,5-di-t-butylcatechol, 3,5-di-tert-butylbenzene-1,2-diol, 3,5-di-tert-butylpyrocatechol, 1,2-benzenediol, 3,5-bis 1,1-dimethylethyl, 4,6-di-tert-butylpyrocatechol, unii-1328kn3f8b, 1,2-benzenediol, 3,5-di 1,1-dimethylethyl, pyrocatechol, 3,5-di-tert-butyl, 3,5-bis tert-butyl benzene-1,2-diol
PubChem CID 66099
IUPAC Name 3,5-ditert-butylbenzene-1,2-diol
SMILES CC(C)(C)C1=CC(=C(C(=C1)O)O)C(C)(C)C

Specifications

Melting Point 95°C to 100°C
Color Brown to White
Quantity 25 g
Beilstein 1370212
Solubility Information Insoluble in water.
Formula Weight 222.33
Percent Purity 99%
Physical Form Solid
Chemical Name or Material 3,5-Di-tert-butylcatechol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 213-816-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.