Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Di-tert-butylcatechol 98.0+%, TCI America™
SDP

Catalog No. D243225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D243225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D243225G Supplier TCI America Supplier No. D243225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1020-31-1
Molecular Formula C14H22O2
Molecular Weight (g/mol) 222.328
MDL Number MFCD00008819
InChI Key PJZLSMMERMMQBJ-UHFFFAOYSA-N
Synonym 3,5-di-tert-butylcatechol, 3,5-di-t-butylcatechol, 3,5-di-tert-butylbenzene-1,2-diol, 3,5-di-tert-butylpyrocatechol, 1,2-benzenediol, 3,5-bis 1,1-dimethylethyl, 4,6-di-tert-butylpyrocatechol, unii-1328kn3f8b, 1,2-benzenediol, 3,5-di 1,1-dimethylethyl, pyrocatechol, 3,5-di-tert-butyl, 3,5-bis tert-butyl benzene-1,2-diol
PubChem CID 66099
IUPAC Name 3,5-ditert-butylbenzene-1,2-diol
SMILES CC(C)(C)C1=CC(=C(C(=C1)O)O)C(C)(C)C

Specifications

Melting Point 99°C
Boiling Point 145°C
Quantity 25 g
Formula Weight 222.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Di-tert-butylcatechol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.