Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Diacetoxybenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. D28445G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28445G 5 g
D28441G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28445G Supplier TCI America Supplier No. D28445G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 35354-29-1
Molecular Formula C11H10O6
Molecular Weight (g/mol) 238.195
MDL Number MFCD00017591
InChI Key QBTDQJMLMVEUTQ-UHFFFAOYSA-N
Synonym 3,5-diacetoxybenzoic acid, 3,5-bis acetyloxy benzoic acid, 3,5-diacethoxybenzoic acid, benzoic acid, 3,5-bis acetyloxy, 3,5-diacetoxy benzoic acid, maybridge1_004330, acmc-209igc, 3.5-diacetoxybenzoic acid, 3,5-diacetyloxybezoic acid, 3,5-diacetoxy-benzoic acid
PubChem CID 539869
IUPAC Name 3,5-diacetyloxybenzoic acid
SMILES CC(=O)OC1=CC(=CC(=C1)C(=O)O)OC(=O)C

Specifications

Melting Point 161°C
Color White-Yellow
Quantity 5 g
Formula Weight 238.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Diacetoxybenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.