Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,5-dibromo-4-methoxybenzoic acid | 4073-35-2 | MFCD00016511 | 309.94 | C8H6Br2O3 | 10 G
SDP

Catalog No. 5000299731
Encompass_Preferred

3,5-dibromo-4-methoxybenzoic acid is a brominated benzoic acid derivative used in chemical and natural-products research. It has been reported as a natural product isolated from marine sponges and is employed as a building block or reference compound in synthetic and analytical applications.

  • Brominated benzoic acid derivative.
  • Reported natural product isolated from marine sponges.
  • Useful as a synthetic building block and reference standard.
  • Molecular formula C8H6Br2O3; molecular weight 309.94.
  • Storage stability: room temperature (3 years); in solvent -80°C (2 years), -20°C (1 year).

Catalog No. 50-002-99731 Supplier Medchemexpress LLC Supplier No. HYW03862710G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3,5-dibromo-4-methoxybenzoic acid is a brominated benzoic acid derivative used in chemical and natural-products research. It has been reported as a natural product isolated from marine sponges and is employed as a building block or reference compound in synthetic and analytical applications.

  • Brominated benzoic acid derivative.
  • Reported natural product isolated from marine sponges.
  • Useful as a synthetic building block and reference standard.
  • Molecular formula C8H6Br2O3; molecular weight 309.94.
  • Storage stability: room temperature (3 years); in solvent -80°C (2 years), -20°C (1 year).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.