Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,5-dibromo-4-methoxybenzoic acid | 4073-35-2 | MFCD00016511 | 309.94 g/mol | C8H6Br2O3 | 25 G
SDP

Catalog No. 5000303025
Encompass_Preferred

3,5-Dibromo-4-methoxybenzoic acid is a brominated aromatic carboxylic acid used in research and organic synthesis. It serves as a building block and intermediate for the preparation of brominated aromatic derivatives and is reported from marine sponge natural products.

  • High chemical purity suitable for research use.
  • Useful building block for synthesis of brominated aromatic compounds.
  • Stable white to off-white solid for convenient handling and storage.
  • Characterized by CAS 4073-35-2 and molecular formula C8H6Br2O3.
  • Available in common laboratory pack sizes for small-scale research.

Catalog No. 50-003-03025 Supplier Medchemexpress LLC Supplier No. 25GHYW038627
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3,5-Dibromo-4-methoxybenzoic acid is a brominated aromatic carboxylic acid used in research and organic synthesis. It serves as a building block and intermediate for the preparation of brominated aromatic derivatives and is reported from marine sponge natural products.

  • High chemical purity suitable for research use.
  • Useful building block for synthesis of brominated aromatic compounds.
  • Stable white to off-white solid for convenient handling and storage.
  • Characterized by CAS 4073-35-2 and molecular formula C8H6Br2O3.
  • Available in common laboratory pack sizes for small-scale research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.