Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,5-dibromo-4-methoxybenzoic acid | 4073-35-2 | MFCD00016511 | >98.0% | 309.94 | C8H6Br2O3 | 5 G
SDP

Catalog No. 5000300529
Encompass_Preferred

3,5-dibromo-4-methoxybenzoic acid is a brominated aromatic carboxylic acid used as a research chemical and reference standard in natural-product and synthetic chemistry. It is found in marine sponges and is used for structure elucidation, as a synthetic intermediate, and in biological studies.

  • CAS number 4073-35-2.
  • Molecular formula C8H6Br2O3.
  • Molecular weight 309.94 g·mol^-1.
  • Synonym 3,5-dibromo-p-anisic acid.
  • Reported purity >98% by some suppliers.
  • Storage stable at room temperature (3 years); solutions store frozen.

Catalog No. 50-003-00529 Supplier Medchemexpress LLC Supplier No. HYW0386275G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3,5-dibromo-4-methoxybenzoic acid is a brominated aromatic carboxylic acid used as a research chemical and reference standard in natural-product and synthetic chemistry. It is found in marine sponges and is used for structure elucidation, as a synthetic intermediate, and in biological studies.

  • CAS number 4073-35-2.
  • Molecular formula C8H6Br2O3.
  • Molecular weight 309.94 g·mol^-1.
  • Synonym 3,5-dibromo-p-anisic acid.
  • Reported purity >98% by some suppliers.
  • Storage stable at room temperature (3 years); solutions store frozen.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.