Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dibromo-L-tyrosine 96.0+%, TCI America™
SDP

Catalog No. D02135G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D02135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D02135G Supplier TCI America Supplier No. D02135G

Chemical Identifiers

CAS 300-38-9
Molecular Formula C9H9Br2NO3
Molecular Weight (g/mol) 338.983
MDL Number MFCD00037204
InChI Key COESHZUDRKCEPA-ZETCQYMHSA-N
PubChem CID 67532
ChEBI CHEBI:28335
IUPAC Name (2S)-2-amino-3-(3,5-dibromo-4-hydroxyphenyl)propanoic acid
SMILES C1=C(C=C(C(=C1Br)O)Br)CC(C(=O)O)N

Specifications

Melting Point 245°C
Color White-Yellow
Quantity 5 g
Formula Weight 338.98
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dibromo-L-tyrosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.