Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dibromobenzotrifluoride, 96%, Thermo Scientific™

Catalog No. AAH6142406
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6142406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6142406 Supplier Thermo Scientific Chemicals Supplier No. H6142406

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3,5-Dibromobenzotrifluoride used as a intermediate in organic synthesis and pharmaceuticals.

Solubility
Slightly soluble in water.

Notes
Recommended storage temperature: 2 - 4°C. Storage: Keep container tightly closed. Store in a cool, dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 401-84-3
Molecular Formula C7H3Br2F3
Molecular Weight (g/mol) 303.90
MDL Number MFCD00526219
InChI Key OFNXSUANJLHGQN-UHFFFAOYSA-N
Synonym 1,3-dibromo-5-trifluoromethyl benzene, 3,5-dibromobenzotrifluoride, benzene, 1,3-dibromo-5-trifluoromethyl, pubchem11234, acmc-209jbn, ksc911m2p, 1,3-dibromobenzotrifluoride, 3,5-dibromo-, ,-trifluorotoluene, 3,5-dibromo-1-trifluoromethyl benzene, 1,3-dibromo-5-trifluoromethyl ;benzene
PubChem CID 2063489
IUPAC Name 1,3-dibromo-5-(trifluoromethyl)benzene
SMILES FC(F)(F)C1=CC(Br)=CC(Br)=C1

Specifications

Color Yellow
Quantity 5 g
Beilstein 2093912
Solubility Information Slightly soluble in water.
Formula Weight 303.9
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 3,5-Dibromobenzotrifluoride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.