Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dibromophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. p-7193903 Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46705G 5 g
D4670-25G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D46705G Supplier TCI America Supplier No. D46705G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material 3,5-Dibromophenylboronic Acid (contains varying amounts of Anhydride)
CAS 117695-55-3
Molecular Formula C6H5BBr2O2
Quantity 5 g
MDL Number MFCD01075725
Synonym 3,5-dibromobenzeneboronic acid, 3,5-dibromophenyl boronic acid, boronic acid, 3,5-dibromophenyl, pubchem14108, acmc-2099ur, amtb596, 3,5-dibromobenzeneboronicacid, ksc493i3d, 3,5-dibromobenzene boronic acid
InChI Key WQBLCGDZYFKINX-UHFFFAOYSA-N
SMILES B(C1=CC(=CC(=C1)Br)Br)(O)O
IUPAC Name (3,5-dibromophenyl)boronic acid
Molecular Weight (g/mol) 279.722
PubChem CID 2734689
Formula Weight 279.72
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.