Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dibromosalicylaldoxime 98.0+%, TCI America™
SDP

Catalog No. D02105G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D02105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D02105G Supplier TCI America Supplier No. D02105G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 21386-43-6
Molecular Formula C7H5Br2NO2
Molecular Weight (g/mol) 294.93
MDL Number MFCD00059607
InChI Key LRYPGXXEEAWIBE-UHFFFAOYSA-N
PubChem CID 19577386
IUPAC Name 2,4-dibromo-6-[(hydroxyamino)methylidene]cyclohexa-2,4-dien-1-one
SMILES ONC=C1C=C(Br)C=C(Br)C1=O

Specifications

Melting Point 227°C
Color White-Yellow
Quantity 5 g
Formula Weight 294.93
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dibromosalicylaldoxime
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.