Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dichloro-2-pyridone, 98+%

Catalog No. AAL0830903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0830903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0830903 Supplier Thermo Scientific Chemicals Supplier No. L0830903

missing translation for 'validMainframeMsg'

CAS: 5437-33-2 | C5H3Cl2NO | 163.985 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5437-33-2
Molecular Formula C5H3Cl2NO
Molecular Weight (g/mol) 163.985
MDL Number MFCD00015401
InChI Key ZICOPWJJZSJEDL-UHFFFAOYSA-N
Synonym 3,5-dichloro-2-pyridone, 3,5-dichloropyridin-2-ol, 3,5-dichloro-2-hydroxypyridine, 2 1h-pyridinone, 3,5-dichloro, 2-pyridinol, 3,5-dichloro, 2 1h-pyridone, 3,5-dichloro, 3,5-dichloro-2-pyridinol, 2-hydroxy-3,5-dichloropyridine, 3,5-dichloropyridin-2 1h-one, acmc-20aiyo
PubChem CID 79496
IUPAC Name 3,5-dichloro-1H-pyridin-2-one
SMILES C1=C(C(=O)NC=C1Cl)Cl

Specifications

Melting Point 178°C to 182°C
Quantity 1 g
Beilstein 1524971
Formula Weight 163.99
Percent Purity ≥98%
Chemical Name or Material 3,5-Dichloro-2-pyridone
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 226-609-3
RTECSNumber UV1146300
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.