Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dichloro-2-pyridone 98.0+%, TCI America™
SDP

Catalog No. D33125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33125G 5 g
D331225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33125G Supplier TCI America Supplier No. D33125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 5437-33-2 | C5H3Cl2NO | 163.985 g/mol

Chemical Identifiers

CAS 5437-33-2
Molecular Formula C5H3Cl2NO
Molecular Weight (g/mol) 163.985
InChI Key ZICOPWJJZSJEDL-UHFFFAOYSA-N
Synonym 3,5-dichloro-2-pyridone, 3,5-dichloropyridin-2-ol, 3,5-dichloro-2-hydroxypyridine, 2 1h-pyridinone, 3,5-dichloro, 2-pyridinol, 3,5-dichloro, 2 1h-pyridone, 3,5-dichloro, 3,5-dichloro-2-pyridinol, 2-hydroxy-3,5-dichloropyridine, 3,5-dichloropyridin-2 1h-one, acmc-20aiyo
PubChem CID 79496
IUPAC Name 3,5-dichloro-1H-pyridin-2-one
SMILES C1=C(C(=O)NC=C1Cl)Cl

Specifications

Melting Point 180°C
Color White-Yellow
Quantity 5 g
Formula Weight 163.99
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dichloro-2-pyridone
RTECSNumber UV1146300
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.