Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3',5'-Dichloro-4'-hydroxyacetophenone 98.0+%, TCI America™
SDP

Catalog No. D50205G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50205G 5 g
D50201G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50205G Supplier TCI America Supplier No. D50205G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17044-70-1
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.034
MDL Number MFCD00016421
InChI Key FXSIZYWHUQEXPC-UHFFFAOYSA-N
PubChem CID 86920
IUPAC Name 1-(3,5-dichloro-4-hydroxyphenyl)ethanone
SMILES CC(=O)C1=CC(=C(C(=C1)Cl)O)Cl

Specifications

Melting Point 164°C
Quantity 5 g
Formula Weight 205.03
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3′,5′-Dichloro-4′-hydroxyacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.