Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,5-dichloro-4-hydroxybenzonitrile | 1891-95-8 | MFCD00002177 | 98.9% | 188.01 g/mol | C7H3Cl2NO | 10 G
SDP

Catalog No. 5000209990
Encompass_Preferred

Chloroxynil (3,5-dichloro-4-hydroxybenzonitrile) is a benzonitrile compound used as a research reagent and as an herbicidal intermediate. It is supplied as a light yellow to yellow solid and is characterized by HPLC purity and spectroscopic verification for laboratory use.

  • Provides the molecular formula C7H3Cl2NO and molecular weight 188.01 g/mol.
  • CAS number 1891-95-8 for unambiguous identification.
  • High purity (HPLC) typically around 98.9% as reported by supplier COA.
  • Appearance: light yellow to yellow solid.
  • Available in solid pack sizes including 10 G.

Catalog No. 50-002-09990 Supplier Medchemexpress LLC Supplier No. HYW05974710G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chloroxynil (3,5-dichloro-4-hydroxybenzonitrile) is a benzonitrile compound used as a research reagent and as an herbicidal intermediate. It is supplied as a light yellow to yellow solid and is characterized by HPLC purity and spectroscopic verification for laboratory use.

  • Provides the molecular formula C7H3Cl2NO and molecular weight 188.01 g/mol.
  • CAS number 1891-95-8 for unambiguous identification.
  • High purity (HPLC) typically around 98.9% as reported by supplier COA.
  • Appearance: light yellow to yellow solid.
  • Available in solid pack sizes including 10 G.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.